Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sick | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273675.1 | NP_001260604.1 | 2 | 1744 | 77 | |
NM_136167.3 | NP_610011.2 | 2 | 1643 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |