Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kua | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201927.2 | NP_001188856.1 | 4 - 5 | 1046 | 67 | |
NM_001273691.1 | NP_001260620.1 | 4 - 5 | 769 | 67 | ||
NM_001273692.1 | NP_001260621.1 | 5 - 6 | 1257 | 67 | ||
NM_136182.2 | NP_610026.1 | 4 - 5 | 750 | 67 | ||
GenBank mRNA | AF152361.1 | - | 3 - 4 | 664 | 67 | |
BT029029.1 | - | 4 - 5 | 750 | 67 | ||
BT126266.1 | - | 5 - 6 | 1257 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |