Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CheB38c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136217.3 | NP_610061.2 | 2 - 3 | 417 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |