Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: step | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273749.1 | NP_001260678.1 | 1 - 2 | 300 | 86 | |
NM_136276.4 | NP_610120.1 | 1 - 2 | 300 | 86 | ||
GenBank mRNA | AY071322.1 | - | 1 - 2 | 234 | 86 | |
BT072836.1 | - | 1 - 2 | 252 | 86 | ||
BT124891.1 | - | 1 - 2 | 199 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |