Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phtf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136388.3 | NP_610232.1 | 7 - 8 | 2445 | 95 | |
GenBank mRNA | AF181638.1 | - | 7 - 8 | 2449 | 95 | |
AJ242938.1 | - | 7 - 8 | 2424 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |