Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgant3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136412.4 | NP_610256.1 | 3 - 4 | 793 | 62 | |
GenBank mRNA | AF145655.1 | - | 3 - 4 | 767 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |