Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kappaB-Ras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144141.3 | NP_001137613.1 | 1 - 2 | 210 | 63 | |
NM_001273841.1 | NP_001260770.1 | 1 - 2 | 210 | 63 | ||
NM_136434.5 | NP_610278.1 | 1 - 2 | 210 | 63 | ||
GenBank mRNA | AY061054.1 | - | 1 - 2 | 155 | 63 |
Gene Symbol: CG30503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144140.3 | NP_001137612.1 | 1 - 2 | 210 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |