Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273852.1 | NP_001260781.1 | 20 - 21 | 6592 | 68 | |
NM_136455.3 | NP_610299.2 | 20 - 21 | 6476 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |