Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CG1339 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136463.2 | NP_610307.2 | 1 - 2 | 1075 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |