Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkc53E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103880.3 | NP_001097350.1 | 12 - 13 | 1922 | 66 | |
NM_001259448.1 | NP_001246377.1 | 7 - 8 | 1253 | 66 | ||
NM_001259449.1 | NP_001246378.1 | 12 - 13 | 1862 | 66 | ||
NM_001274106.1 | NP_001261035.1 | 11 - 12 | 1921 | 66 | ||
NM_057334.3 | NP_476682.1 | 11 - 12 | 2404 | 66 | ||
NM_166201.3 | NP_725626.1 | 12 - 13 | 1925 | 66 | ||
GenBank mRNA | AY095003.1 | - | 12 - 13 | 1896 | 66 | |
BT120171.1 | - | 7 - 8 | 1253 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |