Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sply | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143775.3 | NP_652032.1 | 4 - 5 | 1426 | 64 | |
NM_166215.2 | NP_725652.1 | 3 - 4 | 1363 | 64 | ||
GenBank mRNA | AJ297394.1 | - | 3 - 4 | 1342 | 64 | |
AY052075.1 | - | 4 - 5 | 1374 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |