Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stau | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169714.2 | NP_001163185.1 | 5 - 6 | 3170 | 84 | |
NM_057403.4 | NP_476751.1 | 5 - 6 | 3173 | 84 | ||
NM_166263.2 | NP_725748.1 | 4 - 5 | 2843 | 84 | ||
GenBank mRNA | BT003228.1 | - | 4 - 5 | 2831 | 84 | |
BT010297.1 | - | 5 - 6 | 3191 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |