Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lolal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169715.2 | NP_001163186.1 | 1 - 2 | 489 | 63 | |
NM_001259473.2 | NP_001246402.1 | 2 - 3 | 387 | 63 | ||
NM_001259474.2 | NP_001246403.1 | 2 - 3 | 421 | 63 | ||
NM_001299635.1 | NP_001286564.1 | 2 - 3 | 374 | 63 | ||
NM_080039.4 | NP_524778.1 | 2 - 3 | 352 | 63 | ||
NM_166270.2 | NP_725756.1 | 2 - 3 | 495 | 63 | ||
NM_166271.3 | NP_725757.1 | 2 - 3 | 448 | 63 | ||
NM_166272.3 | NP_725758.1 | 2 - 3 | 412 | 63 | ||
GenBank mRNA | AF308476.1 | - | 2 - 3 | 310 | 63 | |
AY060364.1 | - | 2 - 3 | 308 | 63 | ||
BT099722.1 | - | 1 - 2 | 2071 | 63 | ||
KX531691.1 | - | 1 - 2 | 103 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |