Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hppy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299675.1 | NP_001286604.1 | 4 - 5 | 1019 | 50 | |
NM_001299676.1 | NP_001286605.1 | 4 - 5 | 1019 | 50 | ||
NM_001299677.1 | NP_001286606.1 | 4 - 5 | 1019 | 50 | ||
NM_137555.3 | NP_611399.2 | 4 - 5 | 1019 | 50 | ||
NM_166335.3 | NP_725863.1 | 4 - 5 | 1019 | 50 | ||
GenBank mRNA | BT150295.1 | - | 4 - 5 | 1003 | 50 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |