Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299679.1 | NP_001286608.1 | 13 - 14 | 5124 | 66 | |
NM_079067.4 | NP_523791.2 | 13 - 14 | 5130 | 66 | ||
NM_166338.3 | NP_725866.1 | 12 - 13 | 2133 | 66 | ||
GenBank mRNA | BT133208.1 | - | 12 - 13 | 2131 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |