Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fem-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166408.2 | NP_725993.1 | 2 - 3 | 195 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |