Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079077.4 | NP_523801.2 | 4 - 5 | 977 | 54 | |
NM_166430.3 | NP_726037.1 | 4 - 5 | 858 | 54 | ||
NM_166431.4 | NP_726038.1 | 4 - 5 | 1026 | 54 | ||
GenBank mRNA | AY051890.1 | - | 4 - 5 | 1009 | 54 | |
BT125829.1 | - | 4 - 5 | 977 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |