Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gyg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169761.4 | NP_001163232.2 | 2 - 3 | 513 | 70 | |
NM_001259517.2 | NP_001246446.1 | 2 - 3 | 487 | 70 | ||
NM_001299737.1 | NP_001286666.1 | 2 - 3 | 487 | 70 | ||
NM_001299738.1 | NP_001286667.1 | 2 - 3 | 487 | 70 | ||
NM_001299739.1 | NP_001286668.1 | 2 - 3 | 487 | 70 | ||
NM_166432.3 | NP_726040.1 | 2 - 3 | 487 | 70 | ||
GenBank mRNA | AY121654.1 | - | 2 - 3 | 483 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |