Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080094.4 | NP_524833.2 | 4 - 5 | 1372 | 65 | |
NM_166446.3 | NP_726065.1 | 4 - 5 | 1372 | 65 | ||
NM_176244.3 | NP_788424.1 | 4 - 5 | 1372 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |