Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299747.1 | NP_001286676.1 | 9 - 10 | 3824 | 70 | |
NM_080094.4 | NP_524833.2 | 8 - 9 | 3719 | 70 | ||
NM_166446.3 | NP_726065.1 | 8 - 9 | 3719 | 70 | ||
NM_176244.3 | NP_788424.1 | 8 - 9 | 3719 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |