Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sara | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079990.3 | NP_524729.2 | 4 - 5 | 2753 | 99 | |
GenBank mRNA | AF239997.1 | - | 4 - 5 | 2730 | 99 | |
AJ310804.1 | - | 4 - 5 | 2758 | 99 | ||
BT058000.1 | - | 4 - 5 | 2761 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |