Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppcdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299762.1 | NP_001286691.1 | 3 - 4 | 787 | 64 | |
NM_166475.3 | NP_726105.1 | 2 - 3 | 734 | 64 | ||
GenBank mRNA | AY102691.1 | - | 2 - 3 | 724 | 64 |
Gene Symbol: CG42496 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169772.2 | NP_001163243.1 | 2 - 3 | 734 | 64 | |
NM_001299763.1 | NP_001286692.1 | 3 - 4 | 787 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |