Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: px | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103947.2 | NP_001097417.1 | 12 - 13 | 2911 | 52 | |
NM_001274206.1 | NP_001261135.1 | 2 - 3 | 2062 | 52 | ||
NM_001274207.1 | NP_001261136.1 | 13 - 14 | 4559 | 52 | ||
NM_001382123.1 | NP_001369111.1 | 13 - 14 | 4539 | 52 | ||
NM_001382124.1 | NP_001369112.1 | 13 - 14 | 4539 | 52 | ||
NM_166535.2 | NP_726208.1 | 13 - 14 | 4539 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |