Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: px | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103947.2 | NP_001097417.1 | 3 - 4 | 713 | 112 | |
NM_001274207.1 | NP_001261136.1 | 4 - 5 | 2361 | 112 | ||
NM_001382123.1 | NP_001369111.1 | 4 - 5 | 2341 | 112 | ||
NM_001382124.1 | NP_001369112.1 | 4 - 5 | 2341 | 112 | ||
NM_166535.2 | NP_726208.1 | 4 - 5 | 2341 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |