Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jbug | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169787.2 | NP_001163258.1 | 6 - 7 | 3819 | 70 | |
NM_001274210.1 | NP_001261139.1 | 13 - 14 | 6873 | 70 | ||
NM_001274211.1 | NP_001261140.1 | 14 - 15 | 7221 | 70 | ||
NM_166546.3 | NP_726234.3 | 13 - 14 | 7260 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |