Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jbug | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169787.2 | NP_001163258.1 | 8 - 9 | 5498 | 79 | |
NM_001274210.1 | NP_001261139.1 | 15 - 16 | 8552 | 79 | ||
NM_001274211.1 | NP_001261140.1 | 16 - 17 | 8900 | 79 | ||
NM_166546.3 | NP_726234.3 | 15 - 16 | 8939 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |