Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inaD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259541.1 | NP_001246470.1 | 3 - 4 | 935 | 67 | |
NM_166566.1 | NP_726260.1 | 3 - 4 | 899 | 67 | ||
GenBank mRNA | BT004473.1 | - | 3 - 4 | 900 | 67 | |
BT120028.1 | - | 3 - 4 | 862 | 67 | ||
U15803.1 | - | 3 - 4 | 777 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |