Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inaD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259541.1 | NP_001246470.1 | 5 - 6 | 1518 | 76 | |
NM_166566.1 | NP_726260.1 | 5 - 6 | 1482 | 76 | ||
GenBank mRNA | BT004473.1 | - | 5 - 6 | 1483 | 76 | |
BT120028.1 | - | 5 - 6 | 1445 | 76 | ||
U15803.1 | - | 5 - 6 | 1360 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |