Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nahoda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259545.2 | NP_001246474.1 | 3 - 4 | 1168 | 61 | |
NM_079091.4 | NP_523815.2 | 3 - 4 | 1168 | 61 | ||
NM_166569.3 | NP_726264.1 | 3 - 4 | 700 | 61 | ||
GenBank mRNA | AY060632.1 | - | 3 - 4 | 1125 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |