Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273316.1 | NP_001260245.1 | 1 - 2 | 667 | 78 | |
NM_001273317.1 | NP_001260246.1 | 1 - 2 | 667 | 78 | ||
NM_078786.3 | NP_523510.1 | 1 - 2 | 667 | 78 | ||
GenBank mRNA | AY071690.1 | - | 1 - 2 | 662 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |