Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201817.1 | NP_001188746.1 | 5 - 6 | 1557 | 68 | |
NM_001201818.1 | NP_001188747.1 | 5 - 6 | 1560 | 68 | ||
NM_001201819.2 | NP_001188748.1 | 5 - 6 | 1557 | 68 | ||
NM_001259020.2 | NP_001245949.1 | 4 - 5 | 1689 | 68 | ||
NM_001273351.1 | NP_001260280.1 | 4 - 5 | 1689 | 68 | ||
NM_078797.4 | NP_523521.3 | 4 - 5 | 1689 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |