Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259024.2 | NP_001245953.1 | 1 - 2 | 569 | 89 | |
NM_001298852.1 | NP_001285781.1 | 1 - 2 | 649 | 89 | ||
NM_078800.4 | NP_523524.2 | 1 - 2 | 569 | 89 | ||
GenBank mRNA | AY069697.1 | - | 1 - 2 | 547 | 89 | |
BT003563.1 | - | 3 - 4 | 1250 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Drosophila melanogaster |