Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032069.2 | NP_001027240.1 | 3 - 4 | 1114 | 57 | |
NM_078817.4 | NP_523541.2 | 3 - 4 | 1114 | 57 | ||
GenBank mRNA | BT010016.1 | - | 3 - 4 | 1113 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |