Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr32a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078819.5 | NP_523543.3 | 1 - 2 | 1302 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Drosophila melanogaster |