Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mre11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078823.4 | NP_523547.1 | 2 - 3 | 1389 | 120 | |
GenBank mRNA | AF132144.1 | - | 2 - 3 | 1355 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |