Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spel1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014482.3 | NP_001014482.2 | 1 - 2 | 122 | 76 | |
NM_001259102.2 | NP_001246031.1 | 1 - 2 | 122 | 76 | ||
NM_001299002.1 | NP_001285931.1 | 1 - 2 | 122 | 76 | ||
NM_078841.5 | NP_523565.2 | 1 - 2 | 122 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |