Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABA-B-R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259104.2 | NP_001246033.1 | 7 - 8 | 2252 | 63 | |
NM_078845.5 | NP_523569.3 | 7 - 8 | 2252 | 63 | ||
GenBank mRNA | BT031099.1 | - | 7 - 8 | 1448 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Drosophila melanogaster |