Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yellow-c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299018.1 | NP_001285947.1 | 1 - 2 | 410 | 73 | |
NM_078846.3 | NP_523570.3 | 1 - 2 | 332 | 73 | ||
GenBank mRNA | AY113601.1 | - | 1 - 2 | 333 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |