Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beat-Ic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299041.1 | NP_001285970.1 | 2 - 3 | 1297 | 65 | |
NM_001299042.1 | NP_001285971.1 | 2 - 3 | 1068 | 65 | ||
NM_078856.4 | NP_523580.1 | 2 - 3 | 1068 | 65 | ||
GenBank mRNA | AF325712.1 | - | 2 - 3 | 1050 | 65 | |
AY122127.1 | - | 2 - 3 | 1044 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |