Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: heix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078857.4 | NP_523581.1 | 1 - 2 | 994 | 71 | |
GenBank mRNA | AF440356.2 | - | 1 - 2 | 688 | 71 | |
AY061093.1 | - | 1 - 2 | 1011 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |