Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acp36DE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299111.2 | NP_001286040.1 | 1 - 2 | 213 | 70 | |
NM_078870.4 | NP_523594.1 | 1 - 2 | 213 | 70 | ||
GenBank mRNA | AY113344.1 | - | 1 - 2 | 211 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |