Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259165.1 | NP_001246094.1 | 5 - 6 | 741 | 57 | |
NM_001273665.1 | NP_001260594.1 | 6 - 7 | 1504 | 57 | ||
NM_001273666.1 | NP_001260595.1 | 5 - 6 | 2008 | 57 | ||
NM_001273667.1 | NP_001260596.1 | 6 - 7 | 1177 | 57 | ||
NM_001273668.1 | NP_001260597.1 | 7 - 8 | 1381 | 57 | ||
NM_078880.3 | NP_523604.2 | 4 - 5 | 695 | 57 | ||
NM_165313.4 | NP_724220.2 | 5 - 6 | 1037 | 57 |
Gene Symbol: CG46244 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316402.1 | NP_001303331.1 | 5 - 6 | 2008 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |