Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itgbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316404.1 | NP_001303333.1 | 2 - 3 | 267 | 65 | |
NM_078884.3 | NP_523608.2 | 2 - 3 | 267 | 65 | ||
GenBank mRNA | BT031252.1 | - | 2 - 3 | 252 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |