Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clumsy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042908.2 | NP_001036373.1 | 8 - 9 | 1998 | 80 | |
NM_078886.3 | NP_523610.2 | 8 - 9 | 1994 | 80 | ||
GenBank mRNA | BT150351.1 | - | 8 - 9 | 1967 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |