Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42Ef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078908.3 | NP_523632.2 | 1 - 2 | 431 | 84 | |
GenBank mRNA | AY070898.1 | - | 1 - 2 | 429 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |