Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42El | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078914.3 | NP_523638.1 | 1 - 2 | 243 | 64 | |
GenBank mRNA | AF274019.1 | - | 1 - 2 | 197 | 64 | |
AY051496.1 | - | 1 - 2 | 197 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |