Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tsp42El | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078914.3 | NP_523638.1 | 4 - 5 | 764 | 75 | |
GenBank mRNA | AF274019.1 | - | 4 - 5 | 718 | 75 | |
AY051496.1 | - | 4 - 5 | 718 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |