Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lbm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273828.1 | NP_001260757.1 | 2 - 3 | 136 | 79 | |
NM_078915.3 | NP_523639.1 | 2 - 3 | 242 | 79 | ||
GenBank mRNA | AY071541.1 | - | 2 - 3 | 237 | 79 | |
U49081.1 | - | 2 - 3 | 227 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |