Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lbm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273828.1 | NP_001260757.1 | 3 - 4 | 302 | 60 | |
NM_078915.3 | NP_523639.1 | 3 - 4 | 408 | 60 | ||
GenBank mRNA | AY071541.1 | - | 3 - 4 | 403 | 60 | |
U49081.1 | - | 3 - 4 | 393 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |