Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Or43a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078923.3 | NP_523647.2 | 5 - 6 | 1052 | 88 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Drosophila melanogaster |